Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700020N01Rik PubMed Links |
1700020N01Rik - RIKEN cDNA 1700020N01 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
