Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1810013L24Rik PubMed Links |
1810013L24Rik - RIKEN cDNA 1810013L24 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081400.3 | Intron | NP_001074869.1 |
