Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700008C04Rik PubMed Links |
| 1700008C04Rik - RIKEN cDNA 1700008C04 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Gm40421 - predicted gene, 40421 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1700008C04Rik
Genomic Map