Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2610301B20Rik PubMed Links |
2610301B20Rik - RIKEN cDNA 2610301B20 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026005.3 | 1072 | UTR 3 | NP_080281.3 |
