Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1110051M20Rik PubMed Links |
| 1110051M20Rik - RIKEN cDNA 1110051M20 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_175123.4 | 1659 | Intron | NP_780332.1 | |||
| Arfgap2 - ADP-ribosylation factor GTPase activating protein 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001166024.1 | 1659 | Missense Mutation | NP_001159496.1 | |||
| NM_023854.2 | 1659 | Missense Mutation | NP_076343.2 | |||

1110051M20Rik
Genomic Map