Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
3110001I22Rik PubMed Links |
| 3110001I22Rik - RIKEN cDNA 3110001I22 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_025653.2 | 1121 | Missense Mutation | NP_079929.1 | |||
| Bfar - bifunctional apoptosis regulator | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001177552.1 | 1121 | Intron | NP_001171023.1 | |||
| NM_025976.5 | 1121 | Intron | NP_080252.1 | |||
| Parn - poly(A)-specific ribonuclease (deadenylation nuclease) | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

3110001I22Rik
Genomic Map