Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700011I03Rik PubMed Links |
1700011I03Rik - RIKEN cDNA 1700011I03 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029290.1 | Intron | NP_083566.1 |
