Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4833422C13Rik PubMed Links |
| 4833422C13Rik - RIKEN cDNA 4833422C13 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Ssbp2 - single-stranded DNA binding protein 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_024186.5 | 2743 | UTR 3 | NP_077148.1 | |||
| NM_024272.4 | 2743 | UTR 3 | NP_077234.2 | |||

4833422C13Rik
Genomic Map