Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4921511I17Rik PubMed Links |
| 4921511I17Rik - RIKEN cDNA 4921511I17 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Fam49a - family with sequence similarity 49, member A | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

4921511I17Rik
Genomic Map