Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2200002J24Rik PubMed Links |
| 2200002J24Rik - RIKEN cDNA 2200002J24 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_026961.2 | 131 | Missense Mutation | NP_081237.1 | |||

2200002J24Rik
Genomic Map