Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4921515E04Rik PubMed Links |
| 4921515E04Rik - RIKEN cDNA 4921515E04 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

4921515E04Rik
Genomic Map