Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4921507L20Rik PubMed Links |
| 4921507L20Rik - RIKEN cDNA 4921507L20 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Alkbh3 - alkB, alkylation repair homolog 3 (E. coli) | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_026944.1 | Intron | NP_081220.1 | ||||

4921507L20Rik
Genomic Map