Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700024P16Rik PubMed Links |
| 1700024P16Rik - RIKEN cDNA 1700024P16 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001162980.1 | Intron | NP_001156452.1 | ||||
| NM_001285952.1 | Intron | NP_001272881.1 | ||||
| NM_001285953.1 | Intron | NP_001272882.1 | ||||

1700024P16Rik
Genomic Map