Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2810404M03Rik PubMed Links |
2810404M03Rik - RIKEN cDNA 2810404M03 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
