Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2210015D19Rik PubMed Links |
2210015D19Rik - RIKEN cDNA 2210015D19 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Dbnl - drebrin-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
