Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930402D18Rik PubMed Links |
4930402D18Rik - RIKEN cDNA 4930402D18 gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
A1bg - alpha-1-B glycoprotein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
