Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
3110045C21Rik PubMed Links |
| 3110045C21Rik - RIKEN cDNA 3110045C21 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Ddr2 - discoidin domain receptor family, member 2 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_022563.2 | Intron | NP_072075.2 | ||||
| Hsd17b7 - hydroxysteroid (17-beta) dehydrogenase 7 | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

3110045C21Rik
Genomic Map