Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1500015L24Rik PubMed Links |
| 1500015L24Rik - RIKEN cDNA 1500015L24 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||

1500015L24Rik
Genomic Map