Assay Details
Gene Family ID: | MIPF0000048, mir-128 |
Mature miRNA Sequence: | UCACAGUGAACCGGUCUCUUUU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Ateles geoffroyi | age-miR-128 | MIMAT0002233 | |
Fugu rubripes | fru-miR-128 | MIMAT0003018 | |
Ophiophagus hannah | oha-miR-128-3p | MIMAT0036679 | |
Pan paniscus | ppa-miR-128 | MIMAT0002234 | |
Rhesus monkey | mml-miR-128a-3p | MIMAT0002229 | |
Saguinus labiatus | sla-miR-128 | MIMAT0002232 | |
View More Rows | |||
Tetraodon nigroviridis | tni-miR-128 | MIMAT0003019 | |
Xenopus tropicalis | xtr-miR-128 | MIMAT0003589 | |
Zebrafish | dre-miR-128-3p | MIMAT0001824 |
Stem-loop Details
Ateles geoffroyi
Fugu rubripes
Ophiophagus hannah
Pan paniscus
Rhesus monkey
Saguinus labiatus
Tetraodon nigroviridis
Xenopus tropicalis
Zebrafish

More Information
Related Products
miRBase Alias:
Human: hsa-miR-128a (v9.2)
Mouse: mmu-miR-128a (v9.2)
Rat: rno-miR-128a (v9.2)
Acinetobacter genomosp. 16BJ: age-miR-128a (v10)
Bos taurus: bta-miR-128 (v11)
Bos taurus: bta-miR-128a (v10)
Danio rerio: dre-miR-128 (v20)
Gallus gallus: gga-miR-128a (v8.1)
Gallus gallus: gga-miR-128b (v8.1)
Macaca mulatta: mml-miR-128a (v19)
Pan troglodytes adenovirus: ptr-miR-128a (v10)
Pan troglodytes adenovirus: ptr-miR-128 (v11)
Pectobacterium parvum: ppa-miR-128a (v10)
Pongo pygmaeus: ppy-miR-128a (v10)
Pongo pygmaeus: ppy-miR-128 (v14)
Sphingomonas laterariae: sla-miR-128a (v10)
Staphylococcus schleiferi: ssc-miR-128 (v14)
Staphylococcus schleiferi: ssc-miR-128a (v10)
Mouse: mmu-miR-128a (v9.2)
Rat: rno-miR-128a (v9.2)
Acinetobacter genomosp. 16BJ: age-miR-128a (v10)
Bos taurus: bta-miR-128 (v11)
Bos taurus: bta-miR-128a (v10)
Danio rerio: dre-miR-128 (v20)
Gallus gallus: gga-miR-128a (v8.1)
Gallus gallus: gga-miR-128b (v8.1)
Macaca mulatta: mml-miR-128a (v19)
Pan troglodytes adenovirus: ptr-miR-128a (v10)
Pan troglodytes adenovirus: ptr-miR-128 (v11)
Pectobacterium parvum: ppa-miR-128a (v10)
Pongo pygmaeus: ppy-miR-128a (v10)
Pongo pygmaeus: ppy-miR-128 (v14)
Sphingomonas laterariae: sla-miR-128a (v10)
Staphylococcus schleiferi: ssc-miR-128 (v14)
Staphylococcus schleiferi: ssc-miR-128a (v10)
