Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | UAAGUGCUUCCAUGUUUUGGUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-302a-3p | MIMAT0000684 | |
| Mouse | mmu-miR-302a-3p | MIMAT0000380 | |
| Callithrix jacchus | cja-miR-302a-3p | MIMAT0039587 | |
| Dasypus novemcinctus | dno-miR-302a-3p | MIMAT0047792 | |
| Pan troglodytes | ptr-miR-302a | MIMAT0008086 | |
| Rabbit | ocu-miR-302a-3p | MIMAT0036321 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-302a-3p | MIMAT0006259 | |
Stem-loop Details
Human
Mouse
Callithrix jacchus
Dasypus novemcinctus
Pan troglodytes
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-302a (v17)
Mouse: mmu-miR-302a (v17)
Mouse: mmu-miR-302 (v9.2)
Macaca mulatta: mml-miR-302a (v19)
Mouse: mmu-miR-302a (v17)
Mouse: mmu-miR-302 (v9.2)
Macaca mulatta: mml-miR-302a (v19)

Genomic Map