Assay Details
Gene Family ID: | MIPF0000071, mir-302 |
Mature miRNA Sequence: | UAAGUGCUUCCAUGUUUUGGUGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-302a-3p | MIMAT0000684 |
| Mouse | mmu-miR-302a-3p | MIMAT0000380 |
| Callithrix jacchus | cja-miR-302a-3p | MIMAT0039587 |
| Dasypus novemcinctus | dno-miR-302a-3p | MIMAT0047792 |
| Pan troglodytes | ptr-miR-302a | MIMAT0008086 |
| Rabbit | ocu-miR-302a-3p | MIMAT0036321 |
| View More Rows | ||
| Rhesus monkey | mml-miR-302a-3p | MIMAT0006259 |
Stem-loop Details
Human
Mouse
Callithrix jacchus
Dasypus novemcinctus
Pan troglodytes
Rabbit
Rhesus monkey

Genomic Map