Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCUUCCUUUUAGAGGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-520b-3p | MIMAT0002843 |
| Pan troglodytes | ptr-miR-520b | MIMAT0008198 |
Stem-loop Details
Human
Pan troglodytes
More Information
Related Products
miRBase Alias:
Human: hsa-miR-520b (v21)
Macaca mulatta: mml-miR-520b (v15)
Macaca mulatta: mml-miR-520e (v15)
Macaca mulatta: mml-miR-520b (v15)
Macaca mulatta: mml-miR-520e (v15)

Genomic Map