Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGUGCAUCUUUUUAGAGGAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-519c-3p | MIMAT0002832 |
| Pan troglodytes | ptr-miR-519c | MIMAT0008194 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map