Assay Details
Gene Family ID: | MIPF0000064, mir-31 |
Mature miRNA Sequence: | UGCUAUGCCAACAUAUUGCCAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-31-3p | MIMAT0004504 |
| Pteropus alecto | pal-miR-31-3p | MIMAT0040137 |
Stem-loop Details
Human
Pteropus alecto

Genomic Map