Assay Details
Gene Family ID: | MIPF0000196, mir-335 |
Mature miRNA Sequence: | UUUUUCAUUAUUGCUCCUGACC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-335-3p | MIMAT0004703 | |
| Mouse | mmu-miR-335-3p | MIMAT0004704 | |
| Dasypus novemcinctus | dno-miR-335-3p | MIMAT0047816 | |
| Goat | chi-miR-335-3p | MIMAT0036148 | |
| Guinea pig | cpo-miR-335-3p | MIMAT0047171 | |
| Pteropus alecto | pal-miR-335-3p | MIMAT0040162 | |
| View More Rows | |||
| Rabbit | ocu-miR-335-3p | MIMAT0048383 | |
| Rhesus monkey | mml-miR-335-3p | MIMAT0026853 | |
Stem-loop Details
Human
Mouse
Dasypus novemcinctus
Goat
Guinea pig
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool B v3.0
Related Products
miRBase Alias:
Human: hsa-miR-335* (v17)

Genomic Map