Assay Details
Amplicon Length: | 106 |
Chromosome Location: | Chr. 7 - 130496416 - 130496416 [-] on 130496416 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-335-3p | MIMAT0004703 |
| Human | hsa-miR-335-5p | MIMAT0000765 |
Stem-loop Details
Human

Genomic Map