Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. 6 - 30741706 - 30741706 [+] on 30741706 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-335-3p | MIMAT0004704 |
| Mouse | mmu-miR-335-5p | MIMAT0000766 |
Stem-loop Details
Mouse

Genomic Map