Assay Details
Gene Family ID: | MIPF0000196, mir-335 |
Mature miRNA Sequence: | UUUUUCAUUAUUGCUCCUGACC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-335-3p | MIMAT0004703 |
| Mouse | mmu-miR-335-3p | MIMAT0004704 |
| Dasypus novemcinctus | dno-miR-335-3p | MIMAT0047816 |
| Goat | chi-miR-335-3p | MIMAT0036148 |
| Guinea pig | cpo-miR-335-3p | MIMAT0047171 |
| Pteropus alecto | pal-miR-335-3p | MIMAT0040162 |
| View More Rows | ||
| Rabbit | ocu-miR-335-3p | MIMAT0048383 |
| Rhesus monkey | mml-miR-335-3p | MIMAT0026853 |
Stem-loop Details
Human
Mouse
Dasypus novemcinctus
Goat
Guinea pig
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map