Assay Details
Gene Family ID: | MIPF0000117, mir-139 |
Mature miRNA Sequence: | UCUACAGUGCACGUGUCUCCAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-139-5p | MIMAT0000656 | |
| Rat | rno-miR-139-5p | MIMAT0000845 | |
| Cricetulus griseus | cgr-miR-139-5p | MIMAT0023765 | |
| Gorilla gorilla | ggo-miR-139 | MIMAT0024106 | |
| Horse | eca-miR-139-5p | MIMAT0012983 | |
| Pig | ssc-miR-139-5p | MIMAT0002159 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-139-5p | MIMAT0015761 | |
| Pteropus alecto | pal-miR-139-5p | MIMAT0039925 | |
| Rhesus monkey | mml-miR-139-5p | MIMAT0006195 | |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Gorilla gorilla
Horse
Pig
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-139-5p (v18)

Genomic Map