Assay Details
Gene Family ID: | MIPF0000117, mir-139 |
Mature miRNA Sequence: | UCUACAGUGCACGUGUCUCCAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-139-5p | MIMAT0000656 |
| Rat | rno-miR-139-5p | MIMAT0000845 |
| Cricetulus griseus | cgr-miR-139-5p | MIMAT0023765 |
| Gorilla gorilla | ggo-miR-139 | MIMAT0024106 |
| Horse | eca-miR-139-5p | MIMAT0012983 |
| Pig | ssc-miR-139-5p | MIMAT0002159 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-139-5p | MIMAT0015761 |
| Pteropus alecto | pal-miR-139-5p | MIMAT0039925 |
| Rhesus monkey | mml-miR-139-5p | MIMAT0006195 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Gorilla gorilla
Horse
Pig
Pongo pygmaeus
Pteropus alecto
Rhesus monkey

Genomic Map