Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AAAGCGCUUCCCUUCAGAGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518e-3p | MIMAT0002861 |
| Gorilla gorilla | ggo-miR-518e-3p | MIMAT0036428 |
| Pan troglodytes | ptr-miR-518e | MIMAT0008190 |
| Pongo pygmaeus | ppy-miR-518e-3p | MIMAT0036451 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
More Information
Related Products
miRBase Alias:
Human: hsa-miR-518e (v17)
Pongo pygmaeus: ppy-miR-518g-3p (v21)
Pongo pygmaeus: ppy-miR-518g-3p (v21)

Genomic Map