Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUAAUUGCGGUUUUUGCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548am-5p | MIMAT0022740 |
| Human | hsa-miR-548c-5p | MIMAT0004806 |
| Human | hsa-miR-548o-5p | MIMAT0022738 |
| Pan troglodytes | ptr-miR-548o | MIMAT0024054 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map