Assay Details
Gene Family ID: | MIPF0000046, mir-101 |
Mature miRNA Sequence: | UCAGUUAUCACAGUGCUGAUGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-101a-5p | MIMAT0004526 |
| Rat | rno-miR-101a-5p | MIMAT0004726 |
| Cricetulus griseus | cgr-miR-101a | MIMAT0041145 |
| Goat | chi-miR-101-5p | MIMAT0035899 |
| Oryzias latipes | ola-miR-101a-5p | MIMAT0022542 |
| Taeniopygia guttata | tgu-miR-101-2-5p | MIMAT0014631 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
Goat
Oryzias latipes
Taeniopygia guttata
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-101a* (v17)
Rat: rno-miR-101a* (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-101-5p (v19)
Trabulsiella guamensis ATCC 49490: tgu-miR-101* (v18)
Rat: rno-miR-101a* (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-101-5p (v19)
Trabulsiella guamensis ATCC 49490: tgu-miR-101* (v18)

Genomic Map