Assay Details
Gene Family ID: | MIPF0000097, mir-338 |
Mature miRNA Sequence: | AACAAUAUCCUGGUGCUGAGUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-338-5p | MIMAT0004701 | |
| Mouse | mmu-miR-338-5p | MIMAT0004647 | |
| Rat | rno-miR-338-5p | MIMAT0004646 | |
| Anolis carolinensis | aca-miR-338-5p | MIMAT0021937 | |
| Columba livia | cli-miR-338b-5p | MIMAT0038620 | |
| Horse | eca-miR-338-5p | MIMAT0013035 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-338-5p | MIMAT0015840 | |
| Rhesus monkey | mml-miR-338-5p | MIMAT0006277 | |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Columba livia
Horse
Pongo pygmaeus
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool B v3.0Megaplex™ RT Primers, Rodent Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Rodent MicroRNA B Cards v3.0TaqMan® Human MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Rat: rno-miR-338* (v18)

Genomic Map