Assay Details
Gene Family ID: | MIPF0000097, mir-338 |
Mature miRNA Sequence: | AACAAUAUCCUGGUGCUGAGUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-338-5p | MIMAT0004701 |
Mouse | mmu-miR-338-5p | MIMAT0004647 |
Rat | rno-miR-338-5p | MIMAT0004646 |
Anolis carolinensis | aca-miR-338-5p | MIMAT0021937 |
Columba livia | cli-miR-338b-5p | MIMAT0038620 |
Horse | eca-miR-338-5p | MIMAT0013035 |
View More Rows | ||
Pongo pygmaeus | ppy-miR-338-5p | MIMAT0015840 |
Rhesus monkey | mml-miR-338-5p | MIMAT0006277 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Columba livia
Horse
Pongo pygmaeus
Rhesus monkey
