Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | UAGCAAAAACUGCAGUUACUUU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548p | MIMAT0005934 |
Callithrix jacchus | cja-miR-548v | MIMAT0039619 |
Pan troglodytes | ptr-miR-548p | MIMAT0008228 |
Stem-loop Details
Human
Callithrix jacchus
Pan troglodytes
