Assay Details
Gene Family ID: | MIPF0000635, mir-1329 |
Mature miRNA Sequence: | UACAGUGAUCAGGUUACGAUGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1329-5p | MIMAT0006798 |
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1329 (v18)

Genomic Map