Assay Details
Gene Family ID: | MIPF0000736, mir-2032 |
Mature miRNA Sequence: | UCGCUGCAACAGUUAGCUAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Nematostella vectensis | nve-miR-2032b-5p | MIMAT0009786 |
Stem-loop Details
Nematostella vectensis
More Information
Related Products
miRBase Alias:
Nematostella vectensis: nve-miR-2032b (v18)

Genomic Map