Assay Details
Gene Family ID: | MIPF0000564, mir-1421 |
Mature miRNA Sequence: | GCUGCAUCACGUGUGACCUCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1421ag-3p | MIMAT0007298 |
| Ornithorhynchus anatinus | oan-miR-1421t-3p | MIMAT0007087 |
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1421ag (v19)
Ochrobactrum anthropi: oan-miR-1421t* (v18)
Ochrobactrum anthropi: oan-miR-1421t* (v18)

Genomic Map