Assay Details
| Mature miRNA Sequence: | CAGAUUUCGGAUUUUUUGUA | 
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Bombyx mori | bmo-miR-2852 | MIMAT0013785 | 
Stem-loop Details
Bombyx mori
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | bmo-miR-2852 | 
|---|---|
| miRBase Accession Number: | MI0012992 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | CAGAUUUCGGAUUUUUUGUA | 
| Species: | Bombyx mori | 
| Product Type: | TaqMan™ MicroRNA Assay | 
| Mature miRNA Sequence: | CAGAUUUCGGAUUUUUUGUA | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Bombyx mori | bmo-miR-2852 | MIMAT0013785 | 
 Back To Top
 Back To Top