Assay Details
Gene Family ID: | MIPF0000158, mir-305 |
Mature miRNA Sequence: | AUUGUACUUCAUCAGGUGCUCU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Culex quinquefasciatus | cqu-miR-305-5p | MIMAT0014382 |
Dinoponera quadriceps | dqu-miR-305-5p | MIMAT0045729 |
Heliconius melpomene | hme-miR-305 | MIMAT0024974 |
Ixodes scapularis | isc-miR-305 | MIMAT0018920 |
Stem-loop Details
Culex quinquefasciatus
Dinoponera quadriceps
Heliconius melpomene
Ixodes scapularis

More Information
Related Products
miRBase Alias:
Culex quinquefasciatus: cqu-miR-305 (v18)
