Assay Details
Gene Family ID: | MIPF0000751, mir-2147 |
Mature miRNA Sequence: | AUUUCGCUGUGAGUGUUCCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-2174 | MIMAT0011275 |
Stem-loop Details
Schmidtea mediterranea

Genomic Map