Assay Details
Mature miRNA Sequence: | UCAUUCUACAGGAUCGAAUCAGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-2165-5p | MIMAT0012157 |
Stem-loop Details
Schmidtea mediterranea
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | sme-miR-2165-5p |
|---|---|
| miRBase Accession Number: | MI0010801 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAUUCUACAGGAUCGAAUCAGU |
| Species: | Schmidtea mediterranea |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UCAUUCUACAGGAUCGAAUCAGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-2165-5p | MIMAT0012157 |