Assay Details
Gene Family ID: | MIPF0001793, mir-2058 |
Mature miRNA Sequence: | GGGUGGGGUCUUGCUUCUUUCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma floridae | bfl-miR-2058-3p | MIMAT0019167 |
Stem-loop Details
Branchiostoma floridae
More Information
Related Products
miRBase Alias:
Branchiostoma floridae: bfl-miR-2058* (v18)

Genomic Map