Assay Details
Gene Family ID: | MIPF0001659, mir-1199 |
Mature miRNA Sequence: | UGCGGCCGGUGCUCAACCUGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1199-3p | MIMAT0031120 |
Pan troglodytes | ptr-miR-1199-3p | MIMAT0031128 |
Pongo pygmaeus | ppy-miR-1199-3p | MIMAT0031122 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
