Assay Details
Mature miRNA Sequence: | AUGUAGGGCGAAAAGCCAUGGG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chicken | gga-miR-135a-3-3p | MIMAT0031038 |
Columba livia | cli-miR-135-3-3p | MIMAT0038517 |
Stem-loop Details
Chicken
Columba livia
