Assay Details
Mature miRNA Sequence: | UCGGUUAUCAUGGUACCGGUGC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Chrysemys picta | cpi-miR-101-2-5p | MIMAT0037718 |
Ophiophagus hannah | oha-miR-101b-5p | MIMAT0036652 |
Taeniopygia guttata | tgu-miR-101-1-5p | MIMAT0031101 |
Stem-loop Details
Chrysemys picta
Ophiophagus hannah
Taeniopygia guttata
