Assay Details
Mature miRNA Sequence: | UUAAGACUUGUAGUGAUGUUUA |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Columba livia | cli-miR-499-5p | MIMAT0038660 |
Taeniopygia guttata | tgu-miR-499-5p | MIMAT0025396 |
Stem-loop Details
Columba livia
Taeniopygia guttata
