Assay Details
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUUUAACU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Eptesicus fuscus | efu-miR-499 | MIMAT0034965 |
Stem-loop Details
Eptesicus fuscus

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | efu-miR-499 |
---|---|
miRBase Accession Number: | MI0028650 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUUUAACU |
Species: | Eptesicus fuscus |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000173, mir-499 |
Mature miRNA Sequence: | UUAAGACUUGCAGUGAUGUUUAACU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Eptesicus fuscus | efu-miR-499 | MIMAT0034965 |