Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GCACAUUACACGGUCGACCUCUU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Eptesicus fuscus | efu-miR-323 | MIMAT0035030 |
Stem-loop Details
Eptesicus fuscus

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | efu-miR-323 |
---|---|
miRBase Accession Number: | MI0028715 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GCACAUUACACGGUCGACCUCUU |
Species: | Eptesicus fuscus |
Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | GCACAUUACACGGUCGACCUCUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Eptesicus fuscus | efu-miR-323 | MIMAT0035030 |